Product Details

SNP ID
rs363352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:117200899 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGCAATGCCAGGGCTAAGCTCTGG[C/T]GGGACCCCCATGGGCCTCTCTGTTA
Phenotype
MIM: 613655
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNK18 PubMed Links

Gene Details

Gene
KCNK18
Gene Name
potassium two pore domain channel subfamily K member 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181840.1 Intron NP_862823.1

View Full Product Details