Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhx-1
Entrez Gene ID
181680
Gene Name
Sodium/hydrogen exchanger
Gene Aliases
CELE_B0395.1
Chromosome Location
Chr.X: 16017121 - 16021776 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078221.7 NP_510622.3 5 - 6 772 105

View Full Product Details