Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
uev-3
Entrez Gene ID
185003
Gene Name
protein-coding
Gene Aliases
CELE_F26H9.7
Chromosome Location
Chr.I: 9310315 - 9312642 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001375057.1 NP_001361859.1 5 - 6 773 152
NM_001375060.3 NP_001362168.1 5 - 6 655 152

View Full Product Details