Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nas-5
Entrez Gene ID
188819
Gene Name
Zinc metalloproteinase nas-5
Gene Aliases
CELE_T23H4.3
Chromosome Location
Chr.I: 9886189 - 9888224 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060215.3 NP_492616.1 3 - 4 287 144

View Full Product Details