Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
met-1
Entrez Gene ID
172026
Gene Name
protein-coding
Gene Aliases
CELE_C43E11.3
Chromosome Location
Chr.I: 4254197 - 4264699 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058939.6 NP_491340.2 11 - 12 3986 84
NM_182042.4 NP_871842.2 11 - 12 3957 84

View Full Product Details