Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
met-1
Entrez Gene ID
172026
Gene Name
protein-coding
Gene Aliases
CELE_C43E11.3
Chromosome Location
Chr.I: 4254197 - 4264699 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058939.6 NP_491340.2 1 - 2 245 64

View Full Product Details