Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cbn-1
Entrez Gene ID
173423
Gene Name
protein-coding
Gene Aliases
CELE_K01A2.11
Chromosome Location
Chr.II: 328564 - 332519 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027055.5 NP_001022226.1 4 - 5 606 108
NM_061310.6 NP_493711.2 4 - 5 600 108
NM_061311.6 NP_493712.1 4 - 5 602 108
NR_132575.1 4 - 5 598 108

View Full Product Details