Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cpn-2
Entrez Gene ID
173424
Gene Name
protein-coding
Gene Aliases
CELE_D1069.2
Chromosome Location
Chr.II: 332836 - 339251 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061312.5 NP_493713.2 2 - 3 243 136

View Full Product Details