Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lat-2
Entrez Gene ID
173771
Gene Name
protein-coding
Gene Aliases
CELE_B0286.2
Chromosome Location
Chr.II: 4359065 - 4368281 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047259.5 NP_001040724.1 2 - 3 351 78
GenBank mRNA AY314772.1 - 2 - 3 334 78

View Full Product Details