Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
iffb-1
Entrez Gene ID
175353
Gene Name
Tr-type G domain-containing protein
Gene Aliases
CELE_Y54F10BM.2
Chromosome Location
Chr.III: 2314436 - 2326911 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065135.8 NP_497536.2 1 - 2 84 87
GenBank mRNA DQ867020.1 - 1 - 2 106 87

View Full Product Details