Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sor-1
Entrez Gene ID
176189
Gene Name
protein-coding
Gene Aliases
CELE_ZK1236.3
Chromosome Location
Chr.III: 8418168 - 8422654 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047429.2 NP_001040894.1 2 - 3 1434 68
NM_001379831.2 NP_001367596.1 2 - 3 1443 68

View Full Product Details