Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pat-2
Entrez Gene ID
176240
Gene Name
Integrin alpha pat-2
Gene Aliases
CELE_F54F2.1
Chromosome Location
Chr.III: 8818899 - 8825302 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066547.4 NP_498948.1 9 - 10 2704 85

View Full Product Details