Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ilys-4
Entrez Gene ID
183853
Gene Name
Invertebrate LYSozyme
Gene Aliases
CELE_C55F2.2
Chromosome Location
Chr.IV: 7895628 - 7897545 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383130.1 2 - 3 300 71

View Full Product Details