Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
let-92
Entrez Gene ID
178117
Gene Name
Serine/threonine-protein phosphatase 2A catalytic subunit
Gene Aliases
CELE_F38H4.9
Chromosome Location
Chr.IV: 11856575 - 11860463 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383211.1 2 - 3 191 122
GenBank mRNA AB108533.1 - 1 - 2 130 122

View Full Product Details