Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- cpsf-3
- Entrez Gene ID
-
178285
- Gene Name
- Cleavage and Polyadenylation Specificity Factor
- Gene Aliases
-
CELE_Y67H2A.1
- Chromosome Location
-
Chr.IV: 13273846 - 13282878 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_070152.6 |
NP_502553.2 |
3 - 4 |
819 |
167 |
View Full Product Details