Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ceh-34
Entrez Gene ID
178919
Gene Name
Homeobox protein ceh-34
Gene Aliases
CELE_C10G8.6
Chromosome Location
Chr.V: 5315496 - 5319577 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072018.8 NP_504419.1 1 - 2 177 69

View Full Product Details