Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-31
Entrez Gene ID
188260
Gene Name
Serpentine receptor class gamma-31
Gene Aliases
CELE_T07H8.5
Chromosome Location
Chr.V: 6950847 - 6953897 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072357.4 NP_504758.1 1 - 2 138 100

View Full Product Details