Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hif-1
Entrez Gene ID
180359
Gene Name
protein-coding
Gene Aliases
CELE_F38A6.3
Chromosome Location
Chr.V: 20780407 - 20785260 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028720.5 NP_001023891.2 4 - 5 775 82
NM_001028723.4 NP_001023894.2 3 - 4 656 82
NM_001313065.3 NP_001299994.1 3 - 4 656 82
NM_001373210.2 NP_001359648.1 4 - 5 775 82
NM_075607.7 NP_508008.4 4 - 5 769 82
GenBank mRNA AF364604.1 - 4 - 5 784 82

View Full Product Details