Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nas-38
Entrez Gene ID
180407
Gene Name
Zinc metalloproteinase nas-38
Gene Aliases
CELE_F57C12.1
Chromosome Location
Chr.X: 558141 - 563708 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373215.1 NP_001359993.1 6 - 7 723 120

View Full Product Details