Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hst-6
Entrez Gene ID
189586
Gene Name
Heparan-sulfate 6-O-sulfotransferase
Gene Aliases
CELE_Y34B4A.3
Chromosome Location
Chr.X: 5275093 - 5278519 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001375033.2 NP_001362017.1 5 - 6 622 63
GenBank mRNA AY081844.1 - 4 - 5 600 63

View Full Product Details