Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
apy-1
Entrez Gene ID
181019
Gene Name
Apyrase apy-1
Gene Aliases
CELE_F08C6.6
Chromosome Location
Chr.X: 7568125 - 7573625 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076882.10 NP_509283.1 9 - 10 2224 98

View Full Product Details