Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rig-1
Entrez Gene ID
181115
Gene Name
protein-coding
Gene Aliases
CELE_K09E2.4
Chromosome Location
Chr.X: 8660619 - 8671822 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001375088.2 NP_001362001.1 12 - 13 2273 71

View Full Product Details