Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
snf-12
Entrez Gene ID
181143
Gene Name
Transporter
Gene Aliases
CELE_T25B6.7
Chromosome Location
Chr.X: 9029467 - 9033955 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077126.3 NP_509527.1 8 - 9 1583 103

View Full Product Details