Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sams-1
Entrez Gene ID
181370
Gene Name
Probable S-adenosylmethionine synthase 1
Gene Aliases
CELE_C49F5.1
Chromosome Location
Chr.X: 11965962 - 11968124 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381113.2 NP_001366882.1 1 - 2 132 120

View Full Product Details