Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-93
Entrez Gene ID
3565280
Gene Name
Seven TM Receptor
Gene Aliases
CELE_F59A1.14
Chromosome Location
Chr.V: 17671700 - 17673443 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028827.1 NP_001023998.1 3 - 4 556 126

View Full Product Details