Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYOC
Entrez Gene ID
490344
Gene Name
myocilin
Gene Aliases
-
Chromosome Location
Chr.7: 27335314 - 27346821 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001048030.3 NP_001041495.2 1 - 2 660 100
GenBank mRNA DQ149972.1 - 1 - 2 660 100
DQ303878.2 - 1 - 2 542 100

View Full Product Details