Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDC34
Entrez Gene ID
476759
Gene Name
cell division cycle 34, ubiqiutin conjugating enzyme
Gene Aliases
-
Chromosome Location
Chr.20: 57960506 - 57967388 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_533965.7 XP_533965.3 4 - 5 519 63

View Full Product Details