Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC3
Entrez Gene ID
607593
Gene Name
coiled-coil domain containing 3
Gene Aliases
-
Chromosome Location
Chr.2: 23477696 - 23593107 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_844332.3 XP_849425.1 - 687 94

View Full Product Details