Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SFRP1
Entrez Gene ID
482844
Gene Name
secreted frizzled related protein 1
Gene Aliases
FrzA
Chromosome Location
Chr.16: 24262451 - 24306162 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003639564.2 XP_003639612.1 - 550 154
GenBank mRNA AB672664.1 - - 494 154

View Full Product Details