Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INTS5
Entrez Gene ID
612216
Gene Name
integrator complex subunit 5
Gene Aliases
-
Chromosome Location
Chr.18: 53998828 - 54004127 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_849952.4 XP_855045.1 1 - 2 157 127

View Full Product Details