Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SFRP1
Entrez Gene ID
482844
Gene Name
secreted frizzled related protein 1
Gene Aliases
FrzA
Chromosome Location
Chr.16: 24262451 - 24306162 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003639564.2 XP_003639612.1 - 715 192

View Full Product Details