Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
CARHSP1
Entrez Gene ID
610212
Gene Name
calcium regulated heat stable protein 1
Gene Aliases
-
Chromosome Location
Chr.6: 33338577 - 33345125 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003434858.3 XP_003434906.1 3 - 3 419 133
XM_022420081.1 XP_022275789.1 3 - 3 376 133

View Full Product Details