Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSKH2
Entrez Gene ID
487033
Gene Name
protein serine kinase H2
Gene Aliases
-
Chromosome Location
Chr.29: 32296457 - 32316319 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_544162.4 XP_544162.2 2 - 3 848 156

View Full Product Details