Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FUNDC2
Entrez Gene ID
613014
Gene Name
FUN14 domain containing 2
Gene Aliases
HCBP6
Chromosome Location
Chr.X: 123046451 - 123064580 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033512.1 NP_001028684.1 3 - 4 391 115

View Full Product Details