Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Isca2
Entrez Gene ID
100771623
Gene Name
iron-sulfur cluster assembly 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615144: 46328 - 47870 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511530.2 XP_003511578.1 3 - 4 328 96
GenBank mRNA JI892606.1 - 3 - 4 316 96
JP043328.1 - 3 - 4 316 96

View Full Product Details