Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hrsp12
Entrez Gene ID
100754697
Gene Name
heat-responsive protein 12
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614029: 955072 - 960741 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007644584.1 XP_007642774.1 2 - 3 332 64
GenBank mRNA JP053670.1 - 2 - 3 332 64

View Full Product Details