Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prdm11
Entrez Gene ID
100761907
Gene Name
PR domain containing 11
Gene Aliases
I79_002850
Chromosome Location
Chr.Un|NW_003613646: 656745 - 705360 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007609832.1 XP_007608022.1 1 - 2 126 80

View Full Product Details