Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pycr2
Entrez Gene ID
100751666
Gene Name
pyrroline-5-carboxylate reductase family, member 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614340: 75080 - 78951 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507229.2 XP_003507277.1 3 - 4 509 72

View Full Product Details