Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Igfbp6
Entrez Gene ID
100751810
Gene Name
insulin-like growth factor binding protein 6
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003617247: 26728 - 30980 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003515025.2 XP_003515073.1 3 - 4 598 59

View Full Product Details