Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc20
Entrez Gene ID
100762897
Gene Name
leucine rich repeat containing 20
Gene Aliases
I79_006045
Chromosome Location
Chr.Un|NW_003613759: 867795 - 960830 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499884.2 XP_003499932.1 3 - 4 380 69

View Full Product Details