Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ELOVL1
Entrez Gene ID
102170380
Gene Name
ELOVL fatty acid elongase 1
Gene Aliases
-
Chromosome Location
Chr.3: 18109073 - 18113877 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287178.1 NP_001274107.1 5 - 6 483 104
XM_005678573.3 XP_005678630.2 6 - 7 640 104
XM_018039364.1 XP_017894853.1 7 - 8 707 104
XM_018039370.1 XP_017894859.1 6 - 7 599 104
XM_018039373.1 XP_017894862.1 7 - 8 677 104
GenBank mRNA JO610506.2 - 2 - 3 920 104
KF549985.1 - 5 - 6 483 104

View Full Product Details