Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GALNT8
Entrez Gene ID
102179079
Gene Name
polypeptide N-acetylgalactosaminyltransferase 8
Gene Aliases
-
Chromosome Location
Chr.5: 104320128 - 104356590 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005680979.3 XP_005681036.2 2 - 3 674 67

View Full Product Details