Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSKH2
Entrez Gene ID
102174226
Gene Name
protein serine kinase H2
Gene Aliases
-
Chromosome Location
Chr.14: 5568488 - 5579009 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018058079.1 XP_017913568.1 1 - 2 658 91

View Full Product Details