Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUDT8
Entrez Gene ID
102184975
Gene Name
nudix hydrolase 8
Gene Aliases
-
Chromosome Location
Chr.29: 46522487 - 46524990 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018043528.1 XP_017899017.1 2 - 3 418 119
GenBank mRNA JO439919.2 - 2 - 3 549 119
JO443090.2 - 2 - 3 549 119

View Full Product Details