Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
USF1
Entrez Gene ID
102174176
Gene Name
upstream transcription factor 1
Gene Aliases
-
Chromosome Location
Chr.3: 111685203 - 111692657 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018046387.1 XP_017901876.1 13 - 14 2480 65
XM_018046390.1 XP_017901879.1 13 - 14 2676 65
XM_018046391.1 XP_017901880.1 13 - 14 2513 65
XM_018046392.1 XP_017901881.1 13 - 14 2670 65
XM_018046393.1 XP_017901882.1 13 - 14 2449 65
XM_018046394.1 XP_017901883.1 13 - 14 2297 65

Gene Symbol
TSTD1
Entrez Gene ID
102175173
Gene Name
thiosulfate sulfurtransferase like domain containing 1
Gene Aliases
-
Chromosome Location
Chr.3: 111685203 - 111686678 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018046395.1 XP_017901884.1 3 - 4 432 65
XM_018046396.1 XP_017901885.1 2 - 3 257 65
GenBank mRNA JO319576.2 - 1 - 2 208 65
KA352009.1 - 3 - 4 580 65

View Full Product Details