Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC3
Entrez Gene ID
102174221
Gene Name
coiled-coil domain containing 3
Gene Aliases
-
Chromosome Location
Chr.13: 16764022 - 16851324 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018057014.1 XP_017912503.1 1 - 2 616 68

View Full Product Details