Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp49a1
Entrez Gene ID
36105
Gene Name
CG18377-PA
Gene Aliases
49a1, CG12894, CG18377, Cyp49A1, Cyp49a1p, Dmel\CG18377, Dmel_CG18377
Chromosome Location
Chr.2R: 10449350 - 10458892 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033524
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259327.2 NP_001246256.1 11 - 12 1913 90
NM_136744.4 NP_610588.2 11 - 12 1673 90
NM_165774.3 NP_724937.1 7 - 8 978 90
NM_206081.2 NP_995803.1 11 - 12 1619 90
GenBank mRNA BT056244.1 - 11 - 12 1665 90

View Full Product Details