Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spt5
Entrez Gene ID
53442
Gene Name
CG7626-PA
Gene Aliases
CG7626, DSIF, Dmel\CG7626, Dmel_CG7626, Dspt5, SPT5, dDSIF, dSpt5, spt5
Chromosome Location
Chr.2R: 19437483 - 19441898 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0040273
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144353.4 NP_652610.1 11 - 12 2788 82
GenBank mRNA AF222864.1 - 11 - 12 2764 82
BT023840.1 - 11 - 12 2764 82

View Full Product Details