Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
emb
Entrez Gene ID
34167
Gene Name
embargoed
Gene Aliases
CG13387, CRM1, Crm1, DCRM1, Dmel\CG13387, Dmel_CG13387, Emb, XPO-1, XPO1, Xpo1, crm1, dCRM1, dXPO1, l(2)k16715
Chromosome Location
Chr.2L: 8403573 - 8408853 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0020497
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316387.1 NP_001303316.1 2 - 3 1863 95
NM_164818.2 NP_723391.2 2 - 3 1863 95
GenBank mRNA AJ249178.1 - 1 - 2 1492 95
AY089558.1 - 2 - 3 1847 95

View Full Product Details