Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- sev
- Entrez Gene ID
-
32039
- Gene Name
- sevenless
- Gene Aliases
-
7LES, 7less, CG18085, CT3980, DROSEV, DROSEV1, DmHD-265, Dmel\CG18085, Dmel_CG18085, HD-265, SEV, Sev, sev(AC)[[14]]
- Chromosome Location
-
Chr.X: 11071441 - 11086299 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003366
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078559.3 |
NP_511114.2 |
11 - 12 |
7595 |
58 |
View Full Product Details